missing translation for 'onlineSavingsMsg'
Learn More

NGAL, clone: 17, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen ABS0481702

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15396291

  • £555.01 / 200┬ÁL


Estimated Shipment
06-02-2018
Add to basket

Description

Description

Reactivity: ABS 048-17-02 binds free porcine NGAL in solution. ABS 048-17-02 can be used as a capture antibody in a sandwich ELISA using biotinylated detection antibody ABS 048-28-02.NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/ml
Specifications

Specifications

NGAL/LCN2
17
200μL purified antibody (1mg/mL) in 0.1M PBS and 15mM sodium azide; pH 7.4
Neutrophil gelatinase-associated lipocalin, HNL; p25, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, Lipocalin-2, Oncogene 24p3
Murine
IgG1, κ
Store at 4° C
Monoclonal
Porcine
Western Blot
Unconjugated
Purified
LCN2
Recombinant pig NGAL adsorbed onto aluminum hydroxide gel.
200μL
Primary
100153501
Videos