Learn More
NGAL Monoclonal Antibody (15), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen ABS0381502
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Specificity: ABS 038-15-02 does not cross-react with NGAL from other mammalian species tested. Reactivity: ABS 038-15-02 binds human NGAL in solution and coated onto a solid phase. ABS 038-15-02 can be used as a capture antibody in a sandwich ELISA using biotinylated detection antibodies HYB 211-02-02, ABS 038-04-02, ABS 038-14-02, ABS 038-23-02 and ABS 038-26-02. Using biotinylated ABS 038-15-02 as a detection antibody, the assay reacts equally with NGAL monomer and homodimer on a mass basis. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
NGAL Monoclonal antibody specifically detects NGAL in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
NGAL | |
Monoclonal | |
1.02 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P80188 | |
LCN2 | |
Recombinant human NGAL adsorbed onto aluminum hydroxide gel. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunocytochemistry, Western Blot | |
15 | |
Unconjugated | |
LCN2 | |
24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; alpha-2-microglobulin-related protein; Alpha-2U globulin-related protein; AW212229; HGNC:6526; HNL; LCN2; lipocalin 2; lipocalin 2 (oncogene 24p3); Lipocalin 24p3; Lipocalin2; Lipocalin-2; migration-stimulating factor inhibitor; MSFI; neutrophil gelatinase-associated lipocalin; NGAL; oncogene 24p3; OTTHUMP00000022240; p25; RP23-161B9.11; secreted inducible protein 24; Siderocalin; Siderocalin LCN2; Sip24; SV-40-induced 24P3 protein; uterocalin | |
Mouse | |
Protein A/G | |
RUO | |
3934 | |
4° C, store in dark | |
Liquid |