missing translation for 'onlineSavingsMsg'
Learn More

NGAL, clone: 15, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen ABS0391502

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15326291

  • £598.00 / 200┬ÁL


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Specificity: ABS 039-15-02 shows no cross-reaction with human NGAL.Reactivity: ABS 039-15-02 binds free rat NGAL in solution. ABS 039-15-02 can be used as a capture antibody in a sandwich ELISA using biotinylated detection antibody ABS 039-32-02. Rat NGAL measures 4-400 pg/mL in an optimized assay.NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/ml
Specifications

Specifications

NGAL/LCN2
Monoclonal
1.08mg/mL
PBS with 15mM sodium azide; pH 7.4
P30152
Neutrophil gelatinase-associated lipocalin, HNL, p25, 25kDa alpha-2-microglobulin-related subunit of MMP-9, Lipocalin-2, Oncogene 24p3
Murine
IgG1, κ
200μL
Antibody
Monoclonal
Rat
ELISA
15
Unconjugated
Lcn2
Purified
LCN2
Recombinant rat NGAL precursor with C-terminal His-tag coupled to carrier .
Protein A
Store at 4° C, store in dark
Primary
170496
Videos