missing translation for 'onlineSavingsMsg'
Learn More

NGAL, clone: 14, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen ABS0381402

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15386281

  • £598.00 / 200┬ÁL


Estimated Shipment
08-02-2018
Add to basket

Description

Description

Specificity: ABS 038-14-02 shows some cross-Specificity with NGAL from pig and dog.Reactivity: ABS 038-14-02 binds human NGAL in solution and coated onto a solid phase. ABS 038-14-02 can be used as a capture antibody in a sandwich ELISA using biotinylated detection antibodies HYB 211-01-02, HYB 211-02-02, ABS 038-04-02, ABS 038-15-02 and ABS 038-26-02. Using biotinylated ABS 038-15-02 as a detection antibody, the assay reacts equally with NGAL monomer and homodimer on a mass basis. With biotinylated ABS 038-14-02 detection antibody, the assay is specific for NGAL homodimer and can be used at sample dilutions giving low levels of total NGAL.NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/ml
Specifications

Specifications

NGAL/LCN2
14
200μL purified antibody (1mg/mL) in 0.1M PBS with 0.5M NaCl and 15mM sodium azide; pH 7.4
Purified
LCN2
Recombinant human NGAL adsorbed onto aluminum hydroxide gel.
200μL
Primary
100153501, 3934, 491320
Western Blot
Unconjugated
P80188
Neutrophil gelatinase-associated lipocalin, HNL; p25, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, Lipocalin-2, Oncogene 24p3
Murine
IgG1, κ
Store at 4° C
Monoclonal
Porcine
Videos