Learn More
NGAL Monoclonal Antibody (14), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen ABS0471402
Code : NEW
Additional Details : Weight : 0.01000kg
Description
ABS 047-14-02 binds free canine NGAL. This antibody shows no cross-reaction with human, mouse ,or rat. ABS 047-14-02 can be used as a capture antibody in a sandwich ELISA using biotinylated detection antibody ABS 047-17-02, giving a detection limit of 5 pg/mL in an unoptimized buffer assay. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
NGAL Monoclonal antibody specifically detects NGAL in Canine samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
NGAL | |
Monoclonal | |
0.99 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
0 | |
LCN2 | |
Recombinant dog NGAL adsorbed onto aluminum hydroxide gel | |
400 μL | |
Primary | |
Canine | |
Antibody | |
IgG2b κ |
ELISA, Immunohistochemistry, Western Blot | |
14 | |
Unconjugated | |
LCN2 | |
24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; alpha-2-microglobulin-related protein; Alpha-2U globulin-related protein; AW212229; HGNC:6526; HNL; LCN2; lipocalin 2; lipocalin 2 (oncogene 24p3); Lipocalin 24p3; Lipocalin2; Lipocalin-2; migration-stimulating factor inhibitor; MSFI; neutrophil gelatinase-associated lipocalin; NGAL; oncogene 24p3; OTTHUMP00000022240; p25; RP23-161B9.11; secreted inducible protein 24; Siderocalin; Siderocalin LCN2; Sip24; SV-40-induced 24P3 protein; uterocalin | |
Mouse | |
Protein A/G | |
RUO | |
491320 | |
4° C, store in dark | |
Liquid |