missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NG2 Monoclonal Antibody (D120.43/D4.11/N143.8/N109.6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 372700
Code : 50
Additional Details : Weight : 0.50000kg
Description
NG2 Monoclonal antibody specifically detects NG2 in Human, Rat samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunoprecipitation.Specifications
NG2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Q00657, Q6UVK1 | |
Cspg4 | |
Purified rat NG2 protein. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (Frozen), Immunoprecipitation | |
D120.43/D4.11/N143.8/N109.6 | |
Unconjugated | |
Cspg4 | |
4732461B14Rik; AN2; AN2 proteoglycan; AN2/NG2 proteoglycan; chondroitin sulfate proteoglycan 4; chondroitin sulfate proteoglycan 4 (melanoma-associated); chondroitin sulfate proteoglycan NG2; CSPG4; HMW MAA; HMW-MAA; HSN tumor-specific antigen; Kiaa4232; MCSP; MCSPG; MEL CSPG; melanoma chondroitin sulfate proteoglycan; Melanoma-associated chondroitin sulfate proteoglycan; MEL-CSPG; membrane-spanning proteoglycan NG2; MSK16; nerve/glial antigen 2; Ng2; Proteoglycan AN2 | |
Mouse | |
Protein A | |
RUO | |
1464, 81651 | |
-20°C | |
Liquid |