Learn More
NFKBIL2 Monoclonal Antibody (GT377), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517292
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (83%), Rat (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NFKBIL2 Monoclonal antibody specifically detects NFKBIL2 in Human samples. It is validated for Western Blot.Specifications
NFKBIL2 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q96HA7 | |
TONSL | |
Recombinant protein fragment contains a sequence corresponding to a region within amino acids 1 and 251 of NFKBIL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot | |
GT377 | |
Unconjugated | |
TONSL | |
DNA repair protein; ikappaBR; I-kappa-B-related protein; IKBR; Inhibitor of kappa B-related protein; NF-kappa-B inhibitor-like protein 2; NFKBIL2; Nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 2; TONSL; tonsoku-like; Tonsoku-like protein; tonsoku-like, DNA repair protein | |
Mouse | |
Protein G | |
RUO | |
4796 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |