Learn More
NF-M Mouse anti-Human, Clone: 327921, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3029
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439845
  • £313.76 / 100µg
Estimated Shipment date
from Supplier 10-07-2020
Description
Description
NF-M Monoclonal antibody MAB3029 Specifically detects NF-M in Human samples. It is validated for Western Blotting
Specifications
Specifications
NF-M
327921
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
NEFM
E. coli-derived recombinant human NF-M Ser2-Ser358 Accession # P07197
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4741
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, KnockDown
LYOPH
Western Blot 1-3 ug/mL, Knockout Validated
P07197
NEF3, NEF3neurofilament medium polypeptide, NEFM, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NFM, NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human NF-M in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) NF-H or rhNF-L is observed.