Learn More
Neurturin Mouse anti-Human, Clone: 126424, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3871
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450296
  • £376.32 / 500µg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Neurturin Monoclonal antibody MAB3871 Specifically detects Neurturin in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications
Specifications
Neurturin
126424
Western Blot 1 ug/mL, ELISA, Blockade of Receptor-ligand Interaction
Q99748
neurturin, NRTN, NTN
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Neurturin in direct ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity with recombinant mouse Neurturin, recombinant human (rh) TGF-alpha , rhTGF-beta 2, rhTGF-beta 1.2, rhTGF-beta 3, rhGDNF, rhLAP, or recombinant rat TGF-beta 5 is observed.
Human
Western Blot, ELISA, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
NRTN
E. coli-derived recombinant human Neurturin Ala96-Val197 Accession # Q99748
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4902
Reconstitute at 0.5 mg/mL in sterile PBS.