Learn More
NTF3, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA118390
Code : LD
Additional Details : Weight : 0.01000kg
Product Code. 11562032
  • £300.00 / 100┬ÁL
Estimated Shipment
20-06-2018
Add to basket
Description
Description
PA1-18390 detects NT3 from human, rat, and chicken samples.PA1-18390 has been successfully used in immunohistochemistry, ELISA, Western blot, and inhibition assay procedures.The PA1-18390 immunogen is recombinant human NT3.Reconstitute with 100 ul of distilled water.
Specifications
Specifications
Neurotrophin-3
Polyclonal
whole serum with no preservative
P25433, P20783, P18280
Neurotrophin-3, Neurotrophic factor, HDNF, Nerve growth factor 2, NGF-2, Ntf3, Ntf-3
Rabbit
IgG
Avoid Freeze/Thaw Cycles, store at -20°C
Primary
4908
Western Blot
Unconjugated
NTF3
Serum
NTF3
Recombinant human NT3.
100μL
Antibody
Polyclonal
Rat
Videos