Learn More
Neuropoietin/NP Rat anti-Mouse, Clone: 321525, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2709
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13429645
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Neuropoietin/NP Monoclonal antibody MAB2709 Specifically detects Neuropoietin/NP in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Neuropoietin/NP
321525
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Rat
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Neuropoietin/NP in direct ELISAs and Western blots.
Mouse
Western Blot
LYOPH
Western Blot 1 ug/mL
P83714
Ctf2
E. coli-derived recombinant mouse Neuropoietin/NP Ala23-Ala204 Accession # P83714
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
100289419
Reconstitute at 0.5 mg/mL in sterile PBS.