Learn More
Neurogenin-2 Mouse anti-Human, Rat, Clone: 7G4, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3314
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450006
  • £288.60 / 100µg
Estimated Shipment
17-08-2020
Description
Description
Neurogenin-2 Monoclonal antibody MAB3314 Specifically detects Neurogenin-2 in Human, Rat samples. It is validated for Immunohistochemistry
Specifications
Specifications
Neurogenin-2
7G4
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Atoh4, BHLHA8, bHLHa8MGC46562, Class A basic helix-loop-helix protein 8, Math4A, NEUROG2, neurogenin 2, neurogenin-2, Neurogenin2, NGN2, NGN-2, Protein atonal homolog 4
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detect human and rat Neurogenin-2.
Human, Rat
Immunohistochemistry
LYOPH
Immunohistochemistry 8-25 ug/mL
Purified
NEUROG2
E. coli-derived recombinant rat Neurogenin-2
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
63973
Reconstitute at 0.5 mg/mL in sterile PBS.