Learn More
Neurogenin-1 Mouse anti-Human, Clone: 309707, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3524
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450116
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 26-03-2020
Description
Description
Neurogenin-1 Monoclonal antibody MAB3524 Specifically detects Neurogenin-1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Neurogenin-1
309707
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
NEUROG1
E. coli-derived recombinant human Neurogenin-1 Ala145-His237 Accession # NP_006152
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4762
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
NP_006152
AKA, BHLHA6, bHLHa6NGN-1, Class A basic helix-loop-helix protein 6, Math4C, NeuroD3, NEUROD3neurogenic differentiation 3, NEUROG1, Neurogenic basic-helix-loop-helix protein, Neurogenic differentiation factor 3, neurogenin 1, neurogenin-1, Neurogenin1, NGN, NGN1
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Neurogenin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Neurogenin-3 is observed.