Learn More
Netrin-G2a Rat anti-Mouse, Clone: 349724, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB27441
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439665
  • £249.49 / 100µg
Estimated Shipment
27-06-2019
Description
Description
Netrin-G2a Monoclonal antibody MAB27441 Specifically detects Netrin-G2a in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
Netrin-G2a
349724
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Netrin-G2a in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Netrin-G1a, rmNetrin-1, or rmNetrin-4 is observed.
Mouse
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
NP_598007
Ntng2
Mouse myeloma cell line NS0-derived recombinant mouse Netrin-G2a Asp18-Arg504 Accession # NP_598007
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
84628
Reconstitute at 0.5 mg/mL in sterile PBS.