Learn More
Netrin-G1a Rat anti-Mouse, Clone: 229722, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1166
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448405
  • £390.72 / 500µg
Estimated Shipment date
from Supplier 13-07-2020
Description
Description
Netrin-G1a Monoclonal antibody MAB1166 Specifically detects Netrin-G1a in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Netrin-G1a
229722
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Netrin-G1a in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant chicken (rch) Netrin-1, recombinant mouse Netrin-1, rchNetrin-2, or recombinant human Netrin-4 is observed.
Mouse
Western Blot
LYOPH
Western Blot 1 ug/mL
Q8R4G0
Ntng1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Netrin-G1a His29-Gly513 Accession # Q8R4G0
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
22854
Reconstitute at 0.5 mg/mL in sterile PBS.