Learn More
Neogenin Rat anti-Mouse, Clone: 221519, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1079
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458315
  • £337.93 / 500µg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
Neogenin Monoclonal antibody MAB1079 Specifically detects Neogenin in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Neogenin
221519
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
NEO1
Mouse myeloma cell line NS0-derived recombinant mouse Neogenin Ala42-Ile1033 (Asp442-Leu461 del) Accession # NP_032710
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4756
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
NP_032710
HsT17534, IGDCC2DKFZp547B146, Immunoglobulin superfamily DCC subclass member 2, NEO, NEO1, neogenin, neogenin (chicken) homolog 1, neogenin 1, neogenin homolog 1, NGN, NGNDKFZp547A066
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Neogenin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse DCC is observed.