missing translation for 'onlineSavingsMsg'
Learn More

Nectin-4 Mouse anti-Human, Clone: 337516, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2659

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459605

  • £279.85 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Nectin-4 Monoclonal antibody MAB2659 Specifically detects Nectin-4 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

Nectin-4/PVRL4
337516
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PVRL4
Mouse myeloma cell line NS0-derived recombinant human Nectin-4, Gly27-Val351, Accession # Q96NY8
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
81607
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, CyTOF
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q96NY8
EDSS1, Ig superfamily receptor LNIR, LNIR, LNIRnectin 4, nectin-4, Nectin4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4, PRR4DKFZp686K05193, PVRL4
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human Nectin-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, does not cross-react with recombinant human Nectin-1, -2, -3, or recombinant mouse Nectin-4.
Human
Videos

For Research Use Only