Learn More
Nectin-3 Mouse anti-Human, Clone: 337407, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3064
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459865
  • £249.49 / 100µg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
Nectin-3 Monoclonal antibody MAB3064 Specifically detects Nectin-3 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Nectin-3/PVRL3
337407
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PVRL3
Mouse myeloma cell line NS0-derived recombinant human Nectin-3 Leu56-Asp400 Accession # Q9NQS3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
25945
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9NQS3
CD113, CD113 antigen, CDw113FLJ90624, DKFZp566B0846, nectin 3, nectin-3, Nectin3, poliovirus receptor-related 3, poliovirus receptor-related protein 3, PPR3, PRR3CD113, PVRL3, PVRR3
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Nectin-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Nectin-1, -2, -4, or recombinant mouse Nectin-3 is observed.