Learn More
MYO1E Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544584
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PKPQPKPKPQ VPQCKALYAY DAQDTDELSF NANDIIDIIK EDPSGWWTGR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
MYO1E Polyclonal antibody specifically detects MYO1E in Human samples. It is validated for Western Blot.Specifications
MYO1E | |
Polyclonal | |
Unconjugated | |
MYO1E | |
2310020N23Rik; 9130023P14Rik; AA407778; FSGS6; HuncM-IC; LOW QUALITY PROTEIN: unconventional myosin-Ie; molecular motor; MYO1C; Myo1e; MYO1E variant protein; myosin heavy chain myr 3; myosin IE; myosin-1e; myosin-IC; myosin-Ie; myosin-Ie; unconventional myosin-Ie; myr 3; Myr3; MYR5; unconventional myosin 1E; Unconventional myosin from rat 5; unconventional myosin-Ie | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4643 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q12965 | |
MYO1E | |
Synthetic peptide directed towards the middle region of human MYO1E | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |