missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIOX Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530650
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: mouse kidney, rat kidney. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MIOX Polyclonal antibody specifically detects MIOX in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
MIOX | |
Polyclonal | |
Unconjugated | |
MIOX | |
0610009I10Rik; AI314022; aldehyde reductase (aldose reductase) like 6; aldehyde reductase (aldose reductase)-like 6; aldehyde reductase 6, renal; aldehyde reductase-like 6; Aldrl6; C85427; Inositol oxygenase; kidney-specific protein 32; KSP32; MI oxygenase; Miox; Myo-inositol oxygenase; renal-specific oxido-reducatse; renal-specific oxidoreductase; renal-specific oxido-reductase; Rsor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
252899, 55586, 56727 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9QXN4, Q9QXN5, Q9UGB7 | |
MIOX | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 204 of Human myo-inositol oxygenase | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |