Learn More
Myeloperoxidase, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA118308
Code : LD
Additional Details : Weight : 0.01000kg
Product Code. 11501982
  • £330.00 / 250µg
Estimated Shipment
20-06-2018
Add to basket
Description
Description
PA1-18308 detects Myeloperoxidase from human samples.PA1-18308 has been successfully used in immunohistochemistry (paraffin and frozen) and Western blot procedures.The PA1-18308 immunogen is Myeloperoxidase isolated from human polymorphonuclear leucocytes.Reconstitute with 250 ul of distilled water.
Specifications
Specifications
Myeloperoxidase
Polyclonal
Unconjugated
MPO
Purified
MPO
Myeloperoxidase isolated from human polymorphonuclear leucocytes.
IgG fraction
Avoid Freeze/Thaw Cycles, store at -20°C
Primary
4353
Western Blot
1mg/mL
Antibody with no preservative
P05164
MPO
Rabbit
IgG
250μg