missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloid Lineage Monoclonal Antibody (OX-82), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517562
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17562 targets Myeloid Lineage in FACS, IHC (F), and WB applications and shows reactivity with Rat samples. The MA5-17562 immunogen is anemic rat bone marrow.
Myeloid Lineage Monoclonal antibody specifically detects Myeloid Lineage in Rat samples. It is validated for Flow Cytometry.Specifications
Myeloid Lineage | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Anemic rat bone marrow | |
50 μg | |
Primary | |
4° C, do not freeze | |
Liquid |
Flow Cytometry | |
OX-82 | |
PE | |
Mouse | |
Protein G | |
RUO | |
Rat | |
Antibody | |
IgG1 |