Learn More
MOG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA519602
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with cow, human and common marmoset based on sequence homology. Store antibody at 4oC for 1-2 weeks. For long-term storage, store at -20oC.
MOG Polyclonal antibody specifically detects MOG in Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
MOG | |
Polyclonal | |
Unconjugated | |
MOG | |
B230317G11Rik; BTN6; BTNL11; DAQB-92E24.2; dystonia 2, torsion (autosomal recessive); MGC26137; MOG; MOG alpha 6; MOG alpha-5; MOG alpha-6; MOG AluA; MOG AluB; MOG Ig AluB; MOG Ig-AluB; MOGIG2; MOGIG-2; myelin oligodendrocyte glycoprotein; myelin-oligodendrocyte glycoprotein; NRCLP7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17441, 24558 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide; pH 7.4 | |
Q61885, Q63345 | |
MOG | |
Synthetic peptide conjugated to KLH derived from within residues 200 to the C-terminus of Rat Myelin oligodendrocyte glycoprotein. | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |