missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myc Monoclonal Antibody (9E10), Alexa Fluor™ 555, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1980A555
Code : NEW
Additional Details : Weight : 0.00900kg
Description
c-Myc Monoclonal antibody specifically detects c-Myc in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
c-Myc | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P01106 | |
MYC | |
Synthetic peptide A(408) E E Q K L I S E E D L L R K R R E Q L K H K L E Q L R N S C A(438) of human c-Myc | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
9E10 | |
Alexa Fluor 555 | |
MYC | |
AU016757; Avian myelocytomatosis viral (v-myc) oncogene homolog; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; mMyc; MRTL; Myc; Myc protein; Myc proto-oncogene protein; MYC proto-oncogene, bHLH transcription factor; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; myelocytomatosis viral oncogene homolog; Niard; Nird; Proto-oncogene c-Myc; RNCMYC; Transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene-like protein | |
Mouse | |
Protein A | |
RUO | |
4609 | |
4° C, do not freeze | |
Liquid |