missing translation for 'onlineSavingsMsg'
Learn More

MuSK Mouse anti-Rat, Clone: 78020, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB562

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461006

  • £353.77 / 500µg


Estimated Shipment
02-10-2018
Add to basket

Description

Description

MuSK Monoclonal antibody MAB562 Specifically detects MuSK in Rat samples. It is validated for Western Blotting
Specifications

Specifications

MuSK
78020
Western Blot 1 ug/mL
Q62838
EC 2.7.10, EC 2.7.10.1, MGC126323, MGC126324, muscle, skeletal, receptor tyrosine kinase, MuSK, skeletal receptor tyrosine-protein kinase
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects rat MuSK in direct ELISAs and Western blots.
Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
MUSK
Mouse myeloma cell line NS0-derived recombinant rat MuSK, Glu22-Thr494 (predicted), Accession # Q62838
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4593
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only