Learn More
MTHFD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530407
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (96%), Rat (95%), Zebrafish (85%), Xenopus laevis (89%), Chicken (85%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MTHFD2 Polyclonal antibody specifically detects MTHFD2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
MTHFD2 | |
Polyclonal | |
Unconjugated | |
MTHFD2 | |
AW558851; Bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase, mitochondrial; Methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase (NAD+ dependent), methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase 2; Mthfd2; NAD-dependent methylene tetrahydrofolate dehydrogenase cyclohydrolase; NAD-dependent methylenetetrahydrofolate dehydrogenase; NMDMC | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10797, 17768, 680308 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.61 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P13995, P18155 | |
MTHFD2 | |
Recombinant fragment corresponding to a region within amino acids 141 and 350 of Human MTHFD2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |