missing translation for 'onlineSavingsMsg'
Learn More

Wnt-8a Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2248

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447434

  • £349.86 / 100µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Wnt-8a Polyclonal antibody AF2248 Specifically detects Wnt-8a in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Wnt-8a
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Q64527
protein Wnt-8a, Protein Wnt-8d, wingless-type MMTV integration site family, member 8A, Wnt8a, WNT8DWNT8d
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Wnt-8a in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Wnt-1, rmWnt-3A, rmWnt-4, rmWnt-5A, and rmWnt-10B is observed.
Mouse
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
WNT8A
E. coli-derived recombinant mouse Wnt-8a peptide Trp23-Glu53, Leu196-Arg273 Accession # Q64527
Immunogen affinity purified
RUO
Primary
7478
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only