missing translation for 'onlineSavingsMsg'
Learn More

Wnt-10b Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2110

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467354

  • £342.51 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Wnt-10b Polyclonal antibody AF2110 Specifically detects Wnt-10b in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Wnt-10b
LYOPH
Western Blot 0.1 ug/mL
P48614
protein Wnt-10b, Protein Wnt-12, SHFM6WNT-10B protein, wingless-type MMTV integration site family, member 10B, Wnt10b, Wnt-12, WNT12
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Wnt-10b in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) Wnt-3a, rmWnt-4, rmWnt-5a, and rmWnt-8a is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
WNT10B
E. coli-derived recombinant mouse Wnt-10b peptide Leu42-Glu82, Arg263-Arg332 Accession # P48614
Immunogen affinity purified
RUO
Primary
7480
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only