missing translation for 'onlineSavingsMsg'
Learn More

VSIG2 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4766

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458894

  • £362.60 / 100µg

Estimated Shipment
10-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

VSIG2 Polyclonal antibody AF4766 Specifically detects VSIG2 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

VSIG2
LYOPH
Western Blot 0.1 ug/mL
Q9Z109
2210413P10Rik, Cortical thymocyte-like protein, CTH, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, CTXL, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse VSIG2 in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) VSIG2 is observed and less than 10% cross-reactivity with rhVSIG3 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
VSIG2
Mouse myeloma cell line NS0-derived recombinant mouse VSIG2 Val25-Ala244 (Glu240Asp) Accession # Q9Z109
Immunogen affinity purified
RUO
Primary
23584
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only