Learn More
Tryptase epsilon/BSSP-4 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2059
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487314
  • £362.00 / 100µg
Estimated Shipment
02-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
Tryptase epsilon/BSSP-4 Polyclonal antibody AF2059 Specifically detects Tryptase epsilon/BSSP-4 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Tryptase epsilon/BSSP-4
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse Tryptase ε/BSSP-4, Ala33-Ser306, Accession No. NP_598492
100μg
Primary
70835
Unconjugated
NP_598492
Goat
IgG
RUO
Polyclonal
Murine
Videos