missing translation for 'onlineSavingsMsg'
Learn More

TRIM5 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3550

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438234

  • £289.09 / 100µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TRIM5 Polyclonal antibody AF3550 Specifically detects TRIM5 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

TRIM5
LYOPH
Western Blot 0.1 ug/mL
AAH38937
EC 6.3.2, EC 6.3.2.-, RING finger protein 88, RNF88, RNF88tripartite motif protein TRIM, TRIM5alpha, tripartite motif containing 5, tripartite motif protein TRIM5, tripartite motif-containing 5, tripartite motif-containing protein 5
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse TRIM5 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human TRIM5 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TRIM5
E. coli-derived recombinant mouse TRIM5 Ala2-Gln249 Accession # AAH38937
Immunogen affinity purified
RUO
Primary
85363
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only