missing translation for 'onlineSavingsMsg'
Learn More

TIM-5 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3374

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438134

  • £289.09 / 100µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TIM-5 Polyclonal antibody AF3374 Specifically detects TIM-5 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

TIM-5
LYOPH
Western Blot 0.1 ug/mL
NP_001156829
Gm12169
Mouse myeloma cell line NS0-derived recombinant mouse TIM-5 Phe22-Gly161 Accession # NP_001156829
Immunogen affinity purified
RUO
Primary
210535
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse TIM-5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant mouse (rm) TIM-6 is observed and less than 1% cross-reactivity with rmTIM-2, rmTIM-3, and rmTIM-4 is observed.
Mouse
Videos

For Research Use Only