Learn More
TIM-4 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2826
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487804
  • £382.00 / 100µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
TIM-4 Polyclonal antibody AF2826 Specifically detects TIM-4 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
TIM-4
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse TIM-4, Ala22-Thr279, Accession No. Q6U7R4
100μg
Primary
276891
Unconjugated
Q6U7R4
Goat
IgG
RUO
Polyclonal
Murine
Videos