missing translation for 'onlineSavingsMsg'
Learn More

THSD1 Sheep anti-Mouse, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4784

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13428904

  • £349.86 / 100µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

THSD1 Polyclonal antibody AF4784 Specifically detects THSD1 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

THSD1
LYOPH
Western Blot 0.1 ug/mL
Q9JM61
thrombospondin type-1 domain-containing protein 1, thrombospondin, type I, domain 1, thrombospondin, type I, domain containing 1,4833423O18Rik, TMTSP, TMTSPMGC74971, Transmembrane molecule with thrombospondin module, UNQ3010
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse THSD1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human THSD1 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
THSD1
Mouse myeloma cell line NS0-derived recombinant mouse THSD1 Glu25-Asn412 Accession # Q9JM61
Immunogen affinity purified
RUO
Primary
55901
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only