missing translation for 'onlineSavingsMsg'
Learn More

Thrombopoietin R/Tpo R Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1317

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486874

  • £377.53 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Thrombopoietin R/Tpo R Polyclonal antibody AF1317 Specifically detects Thrombopoietin R/Tpo R in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Thrombopoietin R/Tpo R
LYOPH
Western Blot 0.1 ug/mL
CAA52031
CD110, CD110 antigen, C-MPL, MPL, MPLV, Myeloproliferative leukemia protein, myeloproliferative leukemia virus oncogene, Proto-oncogene c-Mpl, thrombopoietin receptor, TpoR, TPO-R, TPORMPLV
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse Thrombopoietin R/Tpo R in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human Tpo R is observed and less than 2% cross-reactivity with recombinant mouse Epo R is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MPL
Mouse myeloma cell line NS0-derived recombinant mouse Thrombopoietin R/Tpo R, Ser18-Trp483, Accession # CAA52031
Immunogen affinity purified
RUO
Primary
4352
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only