Learn More
TCAM-1 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3546
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488224
  • £362.00 / 100µg
Estimated Shipment
02-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
TCAM-1 Polyclonal antibody AF3546 Specifically detects TCAM-1 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
TCAM-1
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse TCAM-1, Ala18-Gly481, Accession No. AAH50746
100μg
Primary
75870
Unconjugated
AAH50746
Goat
IgG
RUO
Polyclonal
Murine
Videos