missing translation for 'onlineSavingsMsg'
Learn More

Syndecan-3 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2734

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487744

  • £377.53 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Syndecan-3 Polyclonal antibody AF2734 Specifically detects Syndecan-3 in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

SDC3
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Q64519
KIAA0468, N-syndecan, SDC3, SDCN, SYND3syndecan proteoglycan 3, syndecan 3, syndecan 3 (N-syndecan), syndecan neural type, syndecan-3, Syndecan3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse Syndecan-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse Syndecan-4 is observed.
Mouse
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SDC3
Mouse myeloma cell line NS0-derived recombinant mouse Syndecan-3, Ala45-Glu384, Accession # Q64519
Immunogen affinity purified
RUO
Primary
9672
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only