Learn More
Sulfatase Modifying Factor 2/SUMF2 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3454
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488174
  • £362.00 / 100µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
Sulfatase Modifying Factor 2/SUMF2 Polyclonal antibody AF3454 Specifically detects Sulfatase Modifying Factor 2/SUMF2 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Sulfatase Modifying Factor 2/SUMF2
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse Sulfatase Modifying Factor 2/SUMF2, Gln34-Leu308, Accession No. Q8BPG6
100μg
Primary
Murine
Unconjugated
Q8BPG6
Goat