missing translation for 'onlineSavingsMsg'
Learn More

Stella/Dppa3 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2566

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467644

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Stella/Dppa3 Polyclonal antibody AF2566 Specifically detects Stella/Dppa3 in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Stella/Dppa3
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Q8QZY3
developmental pluripotency associated 3, DPPA3, DPPA3 developmental pluripotency associated 3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Stella/Dppa3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human Stella is observed.
Mouse
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
DPPA3
E. coli-derived recombinant mouse Stella/Dppa3 Met1-Asn150 Accession # Q8QZY3
Antigen Affinity-purified
RUO
Primary
359787
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only