missing translation for 'onlineSavingsMsg'
Learn More

Stanniocalcin 2/STC-2 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2837

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487814

  • £377.53 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Stanniocalcin 2/STC-2 Polyclonal antibody AF2837 Specifically detects Stanniocalcin 2/STC-2 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Stanniocalcin 2/STC-2
LYOPH
Western Blot 0.1 ug/mL
O88452
stanniocalcin 2, stanniocalcin-2, STC2, STC-2, STC-2Stanniocalcin-related protein, STC-related protein, STCRP
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse Stanniocalcin 2/STC-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human STC-2 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
STC2
Mouse myeloma cell line NS0-derived recombinant mouse Stanniocalcin 2/STC-2, Thr25-Arg296, Accession # O88452
Immunogen affinity purified
RUO
Primary
8614
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only