missing translation for 'onlineSavingsMsg'
Learn More

Siglec-F Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1706

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467104

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 26-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Siglec-F Polyclonal antibody AF1706 Specifically detects Siglec-F in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Siglec-F
LYOPH
Western Blot 0.1 ug/mL
Q920G3
SAF2, sialic acid binding Ig like lectin 8, SIGLEC-8, SIGLEC8L, SiglecF
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Siglec-F in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Siglec-2, rmSiglec-3, recombinant human (rh) Siglec-5, rhSiglec-7, rhSiglec-9, and rhSiglec-10 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Siglecf
Mouse myeloma cell line NS0-derived recombinant mouse Siglec-F Asp18-Thr437 Accession # Q920G3
Immunogen affinity purified
RUO
Primary
27181
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only