missing translation for 'onlineSavingsMsg'
Learn More

sFRP-2 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1169

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13446784

  • £342.51 / 100µg

Estimated Shipment date
from Supplier 24-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

sFRP-2 Polyclonal antibody AF1169 Specifically detects sFRP-2 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

sFRP-2
LYOPH
Western Blot 0.1 ug/mL
AAB70795
FRP2, FRP-2secreted apoptosis related protein 1, SARP-1, SARP1sFRP-2, SDF-5, Secreted apoptosis-related protein 1, secreted frizzled-related protein 2, sFRP2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse sFRP-2 in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant human SARP2 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SFRP2
Mouse myeloma cell line NS0-derived recombinant mouse sFRP-2 (R&D Systems, Catalog # 1169-FR) Leu25-Cys295 Accession # AAB70795
Immunogen affinity purified
RUO
Primary
6423
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only