missing translation for 'onlineSavingsMsg'
Learn More

Serpin E2/PN1 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2175

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487384

  • £377.53 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Serpin E2/PN1 Polyclonal antibody AF2175 Specifically detects Serpin E2/PN1 in Mouse samples. It is validated for Immunoprecipitation, Neutralization, Western Blotting
Specifications

Specifications

Serpin E2/PN1
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1.1 ug/mL
Q07235
GDN, GdNPF, GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Proteinase Nexin 1, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse Serpin E2/PN1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A4, rhSerpin I2, recombinant mouse (rm) Serpin C1, rmSerpin D1, and rmSerpin F2 is observed.
Mouse
Western Blot, Immunoprecipitation, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SERPINE2
Mouse myeloma cell line NS0-derived recombinant mouse Serpin E2/PN1, Ser20-Pro397, Accession # Q07235
Immunogen affinity purified
RUO
Primary
5270
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only