missing translation for 'onlineSavingsMsg'
Learn More

Serpin D1/Heparin Cofactor II Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1265

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486844

  • £377.53 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Serpin D1/Heparin Cofactor II Polyclonal antibody AF1265 Specifically detects Serpin D1/Heparin Cofactor II in Mouse samples. It is validated for Immunoprecipitation, Neutralization, Western Blotting
Specifications

Specifications

Serpin D1/Heparin Cofactor II
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1.23 ug/mL
P49182
HC2, HC2LS2, HCF2, HCF2clade D (heparin cofactor), member 1, HC-II, Heparin cofactor II, HLS2HCII, leuserpin 2, LS2, Protease inhibitor leuserpin-2, Serpin D1, serpin peptidase inhibitor, clade D (heparin cofactor), member 1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse Serpin D1/Heparin Cofactor II in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A5, recombinant mouse (rm) Serpin C1, and rmSerpin F2 is observed.
Mouse
Western Blot, Immunoprecipitation, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SERPIND1
Mouse myeloma cell line NS0-derived recombinant mouse Serpin D1/Heparin Cofactor II, Glu24-Ser478, Accession # P49182
Immunogen affinity purified
RUO
Primary
3053
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only