missing translation for 'onlineSavingsMsg'
Learn More

SCARA5 Sheep anti-Mouse, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4754

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488884

  • £347.17 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

SCARA5 Polyclonal antibody AF4754 Specifically detects SCARA5 in Mouse samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

SCARA5
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q8K299
FLJ23907, MGC45780, scavenger receptor class A, member 5 (putative), Tesr, testis expressed scavenger receptor, UNQ2938/PRO28700
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse SCARA5 in direct ELISAs and Western blots. In direct ELISAs, less than 40% cross-reactivity with recombinant human SCARA5 is observed.
Mouse
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SCARA5
Chinese hamster ovary cell line CHO-derived recombinant mouse SCARA5, Arg83-Pro491, Accession # Q8K299
Immunogen affinity purified
RUO
Primary
286133
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only