Learn More
Reg2 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2035
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487294
  • £382.00 / 100µg
Estimated Shipment
03-07-2018
Add to basket
Description
Description
Reg2 Polyclonal antibody AF2035 Specifically detects Reg2 in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
Reg2
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
E. coli-derived recombinant mouse Reg2, Asp35-Ala173, Accession No. Q08731
100μg
Primary
19693
Unconjugated
Q08731
Goat
IgG
RUO
Polyclonal
Murine