Learn More
Reg1 Sheep anti-Mouse, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1657
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487074
  • £382.00 / 100µg
Estimated Shipment
05-07-2018
Add to basket
Description
Description
Reg1 Polyclonal antibody AF1657 Specifically detects Reg1 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Reg1
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
E. coli-derived recombinant mouse Reg1 , Gln22-Gly165, Accession No. P43137
100μg
Primary
19692
Unconjugated
P43137
Sheep
IgG
RUO
Polyclonal
Murine