Learn More
TSC22 Goat anti-Mouse, Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3779
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13428364
  • £360.15 / 100µg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
TSC22 Polyclonal antibody AF3779 Specifically detects TSC22 in Mouse, Rat samples. It is validated for Western Blotting
Specifications
Specifications
TSC22/TSC22D1
LYOPH
Western Blot 0.1 ug/mL
P62500.2
Cerebral protein 2, Egr5, KIAA1994, MGC17597, Regulatory protein TSC-22, RP11-269C23.2, TGFB1I4, TGFB-stimulated clone 22 homolog, transforming growth factor beta 1 induced transcript 4, Transforming growth factor beta-1-induced transcript 4 protein, transforming growth factor beta-stimulated protein TSC-22, TSC22 domain family 1, TSC22 domain family protein 1, TSC22 domain family, member 1, TSC22D1, TSC22DKFZp686O19206
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse and rat TSC22 in direct ELISAs and Western blots.
Mouse, Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TSC22D1
E. coli-derived recombinant mouse TSC22 Met1-Ala143 Accession # P62500.2
Immunogen affinity purified
RUO
Primary
8848
Reconstitute at 0.2 mg/mL in sterile PBS.