missing translation for 'onlineSavingsMsg'
Learn More

R-Spondin 4 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4106

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488554

  • £377.53 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

R-Spondin 4 Polyclonal antibody AF4106 Specifically detects R-Spondin 4 in Mouse samples. It is validated for Neutralization
Specifications

Specifications

R-Spondin 4
LYOPH
Neutralization 5.0-20 ug/mL
Q8BJ73
C20orf182, chromosome 20 open reading frame 182, Cristin 4, CRISTIN4, dJ824F16.3, FLJ16018, hRspo4, Roof plate-specific spondin-4, RSPO4, RSpondin 4, R-spondin family, member 4, R-spondin-4
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse R-Spondin 4 in direct ELISAs.
Mouse
Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
RSPO4
E. coli-derived recombinant mouse R-Spondin 4, Tyr21-Pro197, Accession # Q8BJ73
Immunogen affinity purified
RUO
Primary
343637
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only