Learn More
Proliferin Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1623
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13457054
  • £446.88 / 100µg
Estimated Shipment date
from Supplier 25-03-2020
Description
Description
Proliferin Polyclonal antibody AF1623 Specifically detects Proliferin in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Proliferin
LYOPH
Western Blot 0.1 ug/mL
P04095
Prl2c2
Mouse myeloma cell line NS0-derived recombinant mouse Proliferin Phe30-Cys224 Accession # P04095
Immunogen affinity purified
RUO
Primary
18811
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Proliferin in direct ELISAs and Western blots.